Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTATTCTGTTATAGCAGCCCAAAC[A/G]GACTTAAATACATACTCTCTCTACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610236 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LNPK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LNPK - lunapark, ER junction formation factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305008.1 | 5050 | UTR 3 | NP_001291937.1 | |||
NM_001305009.1 | 5050 | UTR 3 | NP_001291938.1 | |||
NM_001305010.1 | 5050 | UTR 3 | NP_001291939.1 | |||
NM_001305011.1 | 5050 | UTR 3 | NP_001291940.1 | |||
NM_030650.2 | 5050 | UTR 3 | NP_085153.1 | |||
XM_005246878.3 | 5050 | UTR 3 | XP_005246935.1 | |||
XM_005246879.4 | 5050 | UTR 3 | XP_005246936.1 | |||
XM_005246880.3 | 5050 | UTR 3 | XP_005246937.1 | |||
XM_006712783.2 | 5050 | UTR 3 | XP_006712846.1 | |||
XM_011511945.2 | 5050 | UTR 3 | XP_011510247.1 | |||
XM_017005055.1 | 5050 | UTR 3 | XP_016860544.1 |