Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCTTTTTGTTCAGTTTTCCCTG[A/C]CTAGAGCACCCATCACAATCTCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102595 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACYP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACYP2 - acylphosphatase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320586.1 | 2585 | Intron | NP_001307515.1 | |||
NM_001320587.1 | 2585 | Intron | NP_001307516.1 | |||
NM_001320588.1 | 2585 | Intron | NP_001307517.1 | |||
NM_001320589.1 | 2585 | Intron | NP_001307518.1 | |||
NM_001320590.1 | 2585 | Intron | NP_001307519.1 | |||
NM_138448.3 | 2585 | Intron | NP_612457.1 | |||
XM_017005406.1 | 2585 | Intron | XP_016860895.1 | |||
XM_017005407.1 | 2585 | Intron | XP_016860896.1 | |||
XM_017005408.1 | 2585 | Intron | XP_016860897.1 | |||
XM_017005409.1 | 2585 | Intron | XP_016860898.1 | |||
XM_017005410.1 | 2585 | Intron | XP_016860899.1 | |||
XM_017005411.1 | 2585 | Intron | XP_016860900.1 | |||
XM_017005412.1 | 2585 | Intron | XP_016860901.1 | |||
XM_017005413.1 | 2585 | Intron | XP_016860902.1 |
TSPYL6 - TSPY like 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003937.2 | 2585 | UTR 3 | NP_001003937.2 |