Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACACCCCACCTCGGGACAACAGA[A/G]CCTCGTGTCTGGGGTGAGGAGAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605452 MIM: 612204 MIM: 613474 | ||||||||||||||||||||
Literature Links: |
ABCB6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCB6 - ATP binding cassette subfamily B member 6 (Langereis blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005689.2 | 2746 | Missense Mutation | GCT,GTT | A,V 810 | NP_005680.1 |
ATG9A - autophagy related 9A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZFAND2B - zinc finger AN1-type containing 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270998.1 | 2746 | Intron | NP_001257927.1 | |||
NM_001270999.1 | 2746 | Intron | NP_001257928.1 | |||
NM_138802.2 | 2746 | Intron | NP_620157.1 | |||
XM_006712285.1 | 2746 | Intron | XP_006712348.1 | |||
XM_006712286.3 | 2746 | Intron | XP_006712349.1 | |||
XM_017003368.1 | 2746 | Intron | XP_016858857.1 |