Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGATCGAGAATGTGATCTAGACC[A/G]TGACCTATTTTTCAAGTTAGAGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600572 | ||||||||||||||||||||
Literature Links: |
SRSF7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SRSF7 - serine and arginine rich splicing factor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031684.2 | 496 | Missense Mutation | CGG,TGG | R,W 131 | NP_001026854.1 | |
NM_001195446.1 | 496 | Missense Mutation | CGG,TGG | R,W 131 | NP_001182375.1 | |
XM_005264484.2 | 496 | Missense Mutation | CGG,TGG | R,W 131 | XP_005264541.1 | |
XM_005264485.2 | 496 | Missense Mutation | CGG,TGG | R,W 131 | XP_005264542.1 | |
XM_011533032.2 | 496 | Missense Mutation | CGG,TGG | R,W 14 | XP_011531334.1 |