Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTTTGAAGACCCCTCCATGAGTT[A/G]CCTTAGTGTGATCTTTAGAGATGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615117 MIM: 610690 | ||||||||||||||||||||
Literature Links: |
C2orf88 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C2orf88 - chromosome 2 open reading frame 88 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIBCH - 3-hydroxyisobutyryl-CoA hydrolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014362.3 | 1232 | Nonsense Mutation | CAA,TAA | Q,* 313 | NP_055177.2 | |
NM_198047.2 | 1232 | Nonsense Mutation | CAA,TAA | Q,* 313 | NP_932164.1 | |
XM_011510953.1 | 1232 | Nonsense Mutation | CAA,TAA | Q,* 313 | XP_011509255.1 | |
XM_011510954.1 | 1232 | Nonsense Mutation | CAA,TAA | Q,* 147 | XP_011509256.1 |