Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACCCAGACCTCGAGTTTGACTC[G/T]CTACAGCCCTGCTTCTACCCGGACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164840 MIM: 605374 MIM: 615968 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYCN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYCN - v-myc avian myelocytomatosis viral oncogene neuroblastoma derived homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293228.1 | 802 | Silent Mutation | TCG,TCT | S,S 23 | NP_001280157.1 | |
NM_001293231.1 | 802 | Intron | NP_001280160.1 | |||
NM_001293233.1 | 802 | UTR 3 | NP_001280162.1 | |||
NM_005378.5 | 802 | Silent Mutation | TCG,TCT | S,S 23 | NP_005369.2 | |
XM_017004168.1 | 802 | Silent Mutation | TCG,TCT | S,S 23 | XP_016859657.1 |
MYCNOS - MYCN opposite strand | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYCNUT - MYCN upstream transcript (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |