Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGATCAGAGTAGAAAAGGTGTT[C/T]GCATTTAAAGGTCTCTGTTTATGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611338 MIM: 612533 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATG4B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATG4B - autophagy related 4B cysteine peptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013325.4 | Intron | NP_037457.3 | ||||
NM_178326.2 | Intron | NP_847896.1 | ||||
XM_005246992.3 | Intron | XP_005247049.2 | ||||
XM_005246993.2 | Intron | XP_005247050.2 | ||||
XM_005246995.3 | Intron | XP_005247052.1 | ||||
XM_005246996.2 | Intron | XP_005247053.1 | ||||
XM_017003637.1 | Intron | XP_016859126.1 | ||||
XM_017003638.1 | Intron | XP_016859127.1 | ||||
XM_017003639.1 | Intron | XP_016859128.1 | ||||
XM_017003640.1 | Intron | XP_016859129.1 |
THAP4 - THAP domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |