Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAATCCGAGGTATTTGAGATCACG[A/G]ACTTCACCACTGCCTCGGAATGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602536 | ||||||||||||||||||||
Literature Links: |
MAP3K19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP3K19 - mitogen-activated protein kinase kinase kinase 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB3GAP1 - RAB3 GTPase activating protein catalytic subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172435.1 | 89 | Missense Mutation | AAC,GAC | N,D 16 | NP_001165906.1 | |
NM_012233.2 | 89 | Missense Mutation | AAC,GAC | N,D 16 | NP_036365.1 | |
XM_011510823.2 | 89 | Missense Mutation | AAC,GAC | N,D 16 | XP_011509125.1 | |
XM_011510825.2 | 89 | Missense Mutation | AAC,GAC | N,D 16 | XP_011509127.1 |