Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGGATAGAAATGATTACCTGGCA[A/T]AACCAATGATAAGGCTGCCAAAGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602736 | ||||||||||||||||||||
Literature Links: |
ATP5G3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP5G3 - ATP synthase, H+ transporting, mitochondrial Fo complex subunit C3 (subunit 9) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002258.4 | 488 | Missense Mutation | TAT,TTT | Y,F 103 | NP_001002258.1 | |
NM_001190329.1 | 488 | Missense Mutation | TAT,TTT | Y,F 103 | NP_001177258.1 | |
NM_001689.4 | 488 | Missense Mutation | TAT,TTT | Y,F 103 | NP_001680.1 |