Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTATAGTGGACTGGGAAACCAAA[G/T]ACCCTGGGGGAGAAAAGGCAGAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602769 | ||||||||||||||||||||
Literature Links: |
DNMT3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNMT3A - DNA methyltransferase 3 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320892.1 | 2802 | Intron | NP_001307821.1 | |||
NM_001320893.1 | 2802 | Silent Mutation | GTA,GTC | V,V 715 | NP_001307822.1 | |
NM_022552.4 | 2802 | Silent Mutation | GTA,GTC | V,V 867 | NP_072046.2 | |
NM_153759.3 | 2802 | Silent Mutation | GTA,GTC | V,V 678 | NP_715640.2 | |
NM_175629.2 | 2802 | Silent Mutation | GTA,GTC | V,V 867 | NP_783328.1 | |
NM_175630.1 | 2802 | Intron | NP_783329.1 | |||
XM_005264175.4 | 2802 | Silent Mutation | GTA,GTC | V,V 867 | XP_005264232.1 | |
XM_005264177.4 | 2802 | Silent Mutation | GTA,GTC | V,V 644 | XP_005264234.1 | |
XM_011532662.2 | 2802 | Silent Mutation | GTA,GTC | V,V 818 | XP_011530964.1 | |
XM_011532663.2 | 2802 | Silent Mutation | GTA,GTC | V,V 812 | XP_011530965.1 | |
XM_011532664.2 | 2802 | Intron | XP_011530966.1 | |||
XM_011532666.2 | 2802 | Silent Mutation | GTA,GTC | V,V 691 | XP_011530968.1 | |
XM_011532667.2 | 2802 | Silent Mutation | GTA,GTC | V,V 644 | XP_011530969.1 | |
XM_017003526.1 | 2802 | Silent Mutation | GTA,GTC | V,V 867 | XP_016859015.1 | |
XM_017003527.1 | 2802 | Silent Mutation | GTA,GTC | V,V 644 | XP_016859016.1 |