Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGAGGCAATCAATCAGCAATGACA[A/C]AAGCGTGGCTACAGTGATGACCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616526 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC38A11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC38A11 - solute carrier family 38 member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199148.1 | 1528 | Missense Mutation | GTG,TTG | V,L 295 | NP_001186077.1 | |
NM_173512.2 | 1528 | Missense Mutation | GTG,TTG | V,L 273 | NP_775783.1 | |
XM_005246350.4 | 1528 | Missense Mutation | GTG,TTG | V,L 351 | XP_005246407.1 | |
XM_006712337.3 | 1528 | Missense Mutation | GTG,TTG | V,L 297 | XP_006712400.1 | |
XM_011510737.2 | 1528 | Missense Mutation | GTG,TTG | V,L 329 | XP_011509039.1 | |
XM_011510742.2 | 1528 | Missense Mutation | GTG,TTG | V,L 156 | XP_011509044.1 | |
XM_017003455.1 | 1528 | Missense Mutation | GTG,TTG | V,L 377 | XP_016858944.1 | |
XM_017003456.1 | 1528 | Missense Mutation | GTG,TTG | V,L 355 | XP_016858945.1 | |
XM_017003457.1 | 1528 | Missense Mutation | GTG,TTG | V,L 352 | XP_016858946.1 | |
XM_017003458.1 | 1528 | Missense Mutation | GTG,TTG | V,L 321 | XP_016858947.1 | |
XM_017003459.1 | 1528 | Missense Mutation | GTG,TTG | V,L 299 | XP_016858948.1 | |
XM_017003460.1 | 1528 | Missense Mutation | GTG,TTG | V,L 377 | XP_016858949.1 | |
XM_017003461.1 | 1528 | Missense Mutation | GTG,TTG | V,L 377 | XP_016858950.1 | |
XM_017003462.1 | 1528 | Missense Mutation | GTG,TTG | V,L 377 | XP_016858951.1 | |
XM_017003463.1 | 1528 | Missense Mutation | GTG,TTG | V,L 229 | XP_016858952.1 | |
XM_017003464.1 | 1528 | Missense Mutation | GTG,TTG | V,L 182 | XP_016858953.1 | |
XM_017003465.1 | 1528 | Missense Mutation | GTG,TTG | V,L 182 | XP_016858954.1 |