Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAGGCTGGCGGGCGCCGGGCCCG[A/G]CGCGGGGGCGCTGCTCATGCCGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614270 MIM: 600836 MIM: 607150 MIM: 611173 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP65 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFAP65 - cilia and flagella associated protein 65 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CRYBA2 - crystallin beta A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_057093.1 | 54 | Missense Mutation | NP_476434.1 | |||
NM_057094.1 | 54 | Missense Mutation | NP_476435.1 |
FEV - FEV, ETS transcription factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100129175 - uncharacterized LOC100129175 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR375 - microRNA 375 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |