Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATAGCCAGACACCCCTGCACCTC[A/G]TCATCCACAGGGACAACACCACCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 617009 | ||||||||||||||||||||
Literature Links: |
ANKRD53 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD53 - ankyrin repeat domain 53 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001115116.1 | 578 | Missense Mutation | ATC,GTC | I,V 181 | NP_001108588.1 | |
NM_024933.3 | 578 | Missense Mutation | ATC,GTC | I,V 181 | NP_079209.3 | |
XM_005264569.4 | 578 | Missense Mutation | ATC,GTC | I,V 147 | XP_005264626.1 | |
XM_011533114.2 | 578 | Missense Mutation | ATC,GTC | I,V 181 | XP_011531416.1 |
TEX261 - testis expressed 261 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |