Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCTGGACACCATTCAGTCCTCCT[C/G]ATAGGGAACGTGGAAGCCCAGCGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607877 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COPS9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COPS9 - COP9 signalosome subunit 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OTOS - otospiralin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_148961.3 | 368 | Missense Mutation | CAG,GAG | Q,E 87 | NP_683764.1 | |
XM_017003409.1 | 368 | Missense Mutation | CAG,GAG | Q,E 106 | XP_016858898.1 | |
XM_017003410.1 | 368 | Missense Mutation | CAG,GAG | Q,E 87 | XP_016858899.1 |