Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTTCCATAGTAAAGAAGAATTA[A/G]GAGATGGACAGGTAAGCATGCACTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615427 | ||||||||||||||||||||
Literature Links: |
ELMOD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ELMOD3 - ELMO domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135021.1 | 380 | Missense Mutation | AGA,GGA | R,G 15 | NP_001128493.1 | |
NM_001135022.1 | 380 | Missense Mutation | AGA,GGA | R,G 15 | NP_001128494.1 | |
NM_001135023.1 | 380 | Missense Mutation | AGA,GGA | R,G 15 | NP_001128495.1 | |
NM_032213.4 | 380 | Missense Mutation | AGA,GGA | R,G 15 | NP_115589.2 | |
XM_005264596.3 | 380 | Missense Mutation | AGA,GGA | R,G 15 | XP_005264653.1 | |
XM_005264597.3 | 380 | Missense Mutation | AGA,GGA | R,G 15 | XP_005264654.1 | |
XM_005264598.3 | 380 | Missense Mutation | AGA,GGA | R,G 15 | XP_005264655.1 | |
XM_017005078.1 | 380 | Missense Mutation | AGA,GGA | R,G 15 | XP_016860567.1 | |
XM_017005079.1 | 380 | Missense Mutation | AGA,GGA | R,G 15 | XP_016860568.1 |
RETSAT - retinol saturase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |