Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTCCTCTTATCTTTACAGTCAC[G/T]GTTGCTGTTATCACATGCAAGTATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605542 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL36G PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL36G - interleukin 36, gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278568.1 | 244 | Silent Mutation | ACG,ACT | T,T 20 | NP_001265497.1 | |
NM_019618.3 | 244 | Silent Mutation | ACG,ACT | T,T 55 | NP_062564.1 |