Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGGTCATAGGGCCTTTGCTTTCC[G/T]GTTCTGTTCGAATTTTTGCCACTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604464 | ||||||||||||||||||||
Literature Links: |
ITSN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITSN2 - intersectin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006277.2 | 5162 | Missense Mutation | CAG,CCG | Q,P 1662 | NP_006268.2 | |
NM_019595.3 | 5162 | Missense Mutation | CAG,CCG | Q,P 1635 | NP_062541.3 | |
NM_147152.2 | 5162 | Intron | NP_671494.2 |