Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCACGCGGTGATCTCCATGAGGGA[C/T]GGAAAACTCACGCTTATGTTCCGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601534 | ||||||||||||||||||||
Literature Links: |
KCNJ3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNJ3 - potassium voltage-gated channel subfamily J member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001260508.1 | 813 | Silent Mutation | GAC,GAT | D,D 206 | NP_001247437.1 | |
NM_001260509.1 | 813 | Silent Mutation | GAC,GAT | D,D 206 | NP_001247438.1 | |
NM_001260510.1 | 813 | Silent Mutation | GAC,GAT | D,D 206 | NP_001247439.1 | |
NM_002239.3 | 813 | Silent Mutation | GAC,GAT | D,D 206 | NP_002230.1 |