Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCTAGGATGCACATGATCTGCC[C/T]GCTCTCCACGTACAAGGAGACATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605459 MIM: 605460 | ||||||||||||||||||||
Literature Links: |
ABCG5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCG5 - ATP binding cassette subfamily G member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022436.2 | 396 | Missense Mutation | AGG,GGG | R,G 79 | NP_071881.1 | |
XM_005264480.3 | 396 | Missense Mutation | AGG,GGG | R,G 79 | XP_005264537.1 | |
XM_006712073.3 | 396 | Missense Mutation | AGG,GGG | R,G 79 | XP_006712136.1 | |
XM_006712074.3 | 396 | Missense Mutation | AGG,GGG | R,G 79 | XP_006712137.1 | |
XM_011533024.2 | 396 | Missense Mutation | AGG,GGG | R,G 79 | XP_011531326.1 | |
XM_011533025.2 | 396 | UTR 5 | XP_011531327.1 | |||
XM_011533026.2 | 396 | Missense Mutation | AGG,GGG | R,G 79 | XP_011531328.1 | |
XM_011533027.2 | 396 | UTR 5 | XP_011531329.1 | |||
XM_011533028.2 | 396 | Intron | XP_011531330.1 |
ABCG8 - ATP binding cassette subfamily G member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022437.2 | 396 | Intron | NP_071882.1 | |||
XM_005264483.3 | 396 | Intron | XP_005264540.1 | |||
XM_011533029.2 | 396 | Intron | XP_011531331.1 | |||
XM_011533030.2 | 396 | Intron | XP_011531332.1 |
DYNC2LI1 - dynein cytoplasmic 2 light intermediate chain 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |