Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGTTCTTTTTTTCAGTTTAGGA[A/G]GATCTGCTTTGGCCTGCTATACAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605238 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNMT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HNMT - histamine N-methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024074.2 | Intron | NP_001019245.1 | ||||
NM_001024075.1 | Intron | NP_001019246.1 | ||||
NM_006895.2 | Intron | NP_008826.1 | ||||
XM_011511064.2 | Intron | XP_011509366.1 | ||||
XM_017003948.1 | Intron | XP_016859437.1 | ||||
XM_017003949.1 | Intron | XP_016859438.1 |