Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTAGCACTCTGTTTCTTTGCTGT[C/G]AACACGATTCTGACGCTGCAGTTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603639 | ||||||||||||||||||||
Literature Links: |
ADAM17 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAM17 - ADAM metallopeptidase domain 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003183.5 | 2655 | Missense Mutation | CAC,GAC | H,D 818 | NP_003174.3 | |
XM_011510375.2 | 2655 | Missense Mutation | CAC,GAC | H,D 787 | XP_011508677.1 | |
XM_011510376.2 | 2655 | Missense Mutation | CAC,GAC | H,D 598 | XP_011508678.1 | |
XM_011510377.2 | 2655 | Missense Mutation | CAC,GAC | H,D 519 | XP_011508679.1 | |
XM_017004785.1 | 2655 | Missense Mutation | CAC,GAC | H,D 519 | XP_016860274.1 | |
XM_017004786.1 | 2655 | Missense Mutation | CAC,GAC | H,D 519 | XP_016860275.1 |
IAH1 - isoamyl acetate-hydrolyzing esterase 1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039613.2 | 2655 | Intron | NP_001034702.1 | |||
NM_001320858.1 | 2655 | Intron | NP_001307787.1 | |||
NM_001320859.1 | 2655 | Intron | NP_001307788.1 | |||
NM_001320860.1 | 2655 | Intron | NP_001307789.1 | |||
NM_001320863.1 | 2655 | Intron | NP_001307792.1 | |||
XM_017003878.1 | 2655 | Intron | XP_016859367.1 |