Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGGCTTAGGCTTAGCAGGGAGC[A/G]AAGGTGATGGAGGTGGTGTCGCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600073 | ||||||||||||||||||||
Literature Links: |
LRP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRP2 - LDL receptor related protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004525.2 | 13938 | Missense Mutation | TCG,TTG | S,L 4618 | NP_004516.2 | |
XM_011511183.2 | 13938 | Missense Mutation | TCG,TTG | S,L 4575 | XP_011509485.1 | |
XM_011511184.2 | 13938 | Missense Mutation | TCG,TTG | S,L 3855 | XP_011509486.1 |