Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGAGCTGGAATAGAGCTGCCCAA[G/T]TGTCTTAATAATGAATGCTCCACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606165 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BOLL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BOLL - boule homolog, RNA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284358.1 | 1530 | UTR 3 | NP_001271287.1 | |||
NM_001284361.1 | 1530 | Missense Mutation | ATT,CTT | I,L 313 | NP_001271290.1 | |
NM_001284362.1 | 1530 | UTR 3 | NP_001271291.1 | |||
NM_033030.5 | 1530 | UTR 3 | NP_149019.1 | |||
NM_197970.2 | 1530 | UTR 3 | NP_932074.1 | |||
XM_006712715.3 | 1530 | UTR 3 | XP_006712778.1 | |||
XM_011511692.2 | 1530 | UTR 3 | XP_011509994.1 | |||
XM_011511693.2 | 1530 | UTR 3 | XP_011509995.1 | |||
XM_011511694.2 | 1530 | UTR 3 | XP_011509996.1 | |||
XM_017004773.1 | 1530 | Intron | XP_016860262.1 |