Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATGTTTCAACTTGCAAGGAAGAA[C/T]GCCCTTATTGAGTTGATAGAACTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601524 | ||||||||||||||||||||
Literature Links: |
GRB14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRB14 - growth factor receptor bound protein 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303422.1 | 1554 | Missense Mutation | ATT,GTT | I,V 439 | NP_001290351.1 | |
NM_004490.2 | 1554 | Missense Mutation | ATT,GTT | I,V 526 | NP_004481.2 | |
XM_011511022.2 | 1554 | Intron | XP_011509324.1 | |||
XM_017003899.1 | 1554 | Missense Mutation | ATT,GTT | I,V 326 | XP_016859388.1 |