Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCAGTGATTCTGAAAATTACAAG[G/T]GGCAGGGTGCAGGACCTTAAGTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137167 MIM: 601468 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GGCX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GGCX - gamma-glutamyl carboxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000821.6 | 2980 | UTR 3 | NP_000812.2 | |||
NM_001142269.3 | 2980 | UTR 3 | NP_001135741.1 | |||
NM_001311312.1 | 2980 | Intron | NP_001298241.1 | |||
XM_005264259.4 | 2980 | UTR 3 | XP_005264316.1 | |||
XM_011532764.2 | 2980 | UTR 3 | XP_011531066.1 | |||
XM_011532765.2 | 2980 | UTR 3 | XP_011531067.1 | |||
XM_017003803.1 | 2980 | UTR 3 | XP_016859292.1 |
MAT2A - methionine adenosyltransferase 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |