Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGAGCTGTTACCTTTGAGAGCTCA[C/T]TTGCTTGCTGGAAGTAATTAGCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 157655 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NDUFS1 - NADH:ubiquinone oxidoreductase core subunit S1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199981.1 | 2109 | Missense Mutation | AAT,AGT | N,S 633 | NP_001186910.1 | |
NM_001199982.1 | 2109 | Missense Mutation | AAT,AGT | N,S 558 | NP_001186911.1 | |
NM_001199983.1 | 2109 | Missense Mutation | AAT,AGT | N,S 612 | NP_001186912.1 | |
NM_001199984.1 | 2109 | Missense Mutation | AAT,AGT | N,S 683 | NP_001186913.1 | |
NM_005006.6 | 2109 | Missense Mutation | AAT,AGT | N,S 669 | NP_004997.4 | |
XM_017004188.1 | 2109 | Missense Mutation | AAT,AGT | N,S 416 | XP_016859677.1 |