Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGCTTCTTTTCTGGAGACAGCT[G/T]GGTCACGTCGAGGAGGGCACCCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608891 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4757 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4757 - microRNA 4757 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OSR1 - odd-skipped related transciption factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145260.2 | 1635 | Missense Mutation | AAG,CAG | K,Q 155 | NP_660303.1 | |
XM_006711942.3 | 1635 | Missense Mutation | AAG,CAG | K,Q 155 | XP_006712005.1 |