Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCAATCCAATCACTCAGGCCCAG[A/C]GCTGGTGGGAGTTCTGGATTGCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611849 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL44 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPL44 - mitochondrial ribosomal protein L44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022915.3 | 556 | Intron | NP_075066.1 | |||
XM_011511668.2 | 556 | Intron | XP_011509970.1 | |||
XM_017004730.1 | 556 | Intron | XP_016860219.1 | |||
XM_017004731.1 | 556 | UTR 5 | XP_016860220.1 |
WDFY1 - WD repeat and FYVE domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020830.4 | 556 | Intron | NP_065881.1 |