Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGGTCAACGAGTCGGAGAACCA[C/G]AGCAGCAGCTCGGACTCGGAGGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604652 | ||||||||||||||||||||
Literature Links: |
TCF7L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TCF7L1 - transcription factor 7 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031283.2 | 453 | Missense Mutation | CAC,CAG | H,Q 76 | NP_112573.1 | |
XM_006712109.2 | 453 | Missense Mutation | CAC,CAG | H,Q 76 | XP_006712172.1 |