Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTATTTTAGCCAAAAAACAGTCCA[C/T]GGAAACCTCAATTAACTATAATTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607306 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRD5A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SRD5A2 - steroid 5 alpha-reductase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000348.3 | 4035 | Intron | NP_000339.2 | |||
XM_011533069.2 | 4035 | UTR 3 | XP_011531371.1 | |||
XM_011533072.2 | 4035 | UTR 3 | XP_011531374.1 |