Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGAGTATGATTATTGAACTCCAC[A/G]GTGTCCCGGAAGTACAGGGCAGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600239 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR1 - G protein-coupled receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098199.1 | 758 | Silent Mutation | ACC,ACT | T,T 178 | NP_001091669.1 | |
NM_001261452.1 | 758 | Silent Mutation | ACC,ACT | T,T 178 | NP_001248381.1 | |
NM_001261453.1 | 758 | Silent Mutation | ACC,ACT | T,T 178 | NP_001248382.1 | |
NM_001261454.1 | 758 | Silent Mutation | ACC,ACT | T,T 178 | NP_001248383.1 | |
NM_001261455.1 | 758 | Silent Mutation | ACC,ACT | T,T 178 | NP_001248384.1 | |
NM_005279.3 | 758 | Silent Mutation | ACC,ACT | T,T 178 | NP_005270.2 | |
XM_005246471.2 | 758 | Silent Mutation | ACC,ACT | T,T 178 | XP_005246528.1 | |
XM_017003832.1 | 758 | Silent Mutation | ACC,ACT | T,T 178 | XP_016859321.1 |