Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGTTTCAGTGGAGAGCAAAATGA[A/G]TAACAAAGCGGGCTCCTTTTTCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612322 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FASTKD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FASTKD2 - FAST kinase domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136193.1 | 478 | Missense Mutation | AAT,AGT | N,S 18 | NP_001129665.1 | |
NM_001136194.1 | 478 | Missense Mutation | AAT,AGT | N,S 18 | NP_001129666.1 | |
NM_014929.3 | 478 | Missense Mutation | AAT,AGT | N,S 18 | NP_055744.2 |
MDH1B - malate dehydrogenase 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |