Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTAATATCTCTTTAGCATAAAGT[A/G]TTCTATGAAGTCCAGCTTCTCGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC148 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC148 - coiled-coil domain containing 148 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301684.1 | 1940 | Missense Mutation | NP_001288613.1 | |||
NM_001301685.1 | 1940 | Intron | NP_001288614.1 | |||
NM_138803.3 | 1940 | Missense Mutation | NP_620158.3 | |||
XM_005246319.3 | 1940 | Missense Mutation | XP_005246376.1 | |||
XM_011510638.2 | 1940 | Missense Mutation | XP_011508940.1 | |||
XM_017003392.1 | 1940 | Missense Mutation | XP_016858881.1 | |||
XM_017003393.1 | 1940 | Missense Mutation | XP_016858882.1 | |||
XM_017003394.1 | 1940 | Missense Mutation | XP_016858883.1 | |||
XM_017003395.1 | 1940 | Missense Mutation | XP_016858884.1 | |||
XM_017003396.1 | 1940 | Missense Mutation | XP_016858885.1 | |||
XM_017003397.1 | 1940 | Missense Mutation | XP_016858886.1 | |||
XM_017003398.1 | 1940 | Missense Mutation | XP_016858887.1 |
CCDC148-AS1 - CCDC148 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |