Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGTAGACAGCTTATCTCCTGTGC[C/T]GCACCCTTCTGATTTGATCCTGACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611479 MIM: 612762 | ||||||||||||||||||||
Literature Links: |
GPN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPN1 - GPN-loop GTPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145047.1 | 1013 | Missense Mutation | CCG,CTG | P,L 305 | NP_001138519.1 | |
NM_001145048.1 | 1013 | Missense Mutation | CCG,CTG | P,L 238 | NP_001138520.1 | |
NM_001145049.1 | 1013 | Missense Mutation | CCG,CTG | P,L 222 | NP_001138521.1 | |
NM_007266.3 | 1013 | Missense Mutation | CCG,CTG | P,L 331 | NP_009197.2 |
SUPT7L - SPT7-like STAGA complex gamma subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282729.1 | 1013 | Intron | NP_001269658.1 | |||
NM_001282730.1 | 1013 | Intron | NP_001269659.1 | |||
NM_001282731.1 | 1013 | Intron | NP_001269660.1 | |||
NM_001282732.1 | 1013 | Intron | NP_001269661.1 | |||
NM_014860.2 | 1013 | Intron | NP_055675.1 | |||
XM_005264672.4 | 1013 | Intron | XP_005264729.1 |