Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCATATCCCTTCTTCAGCAACCCA[C/T]ATTGCCAGATCACATCAGCACACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604155 | ||||||||||||||||||||
Literature Links: |
LANCL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LANCL1 - LanC like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136574.1 | 1010 | Missense Mutation | TAT,TGT | Y,C 312 | NP_001130046.1 | |
NM_001136575.1 | 1010 | Missense Mutation | TAT,TGT | Y,C 312 | NP_001130047.1 | |
NM_006055.2 | 1010 | Missense Mutation | TAT,TGT | Y,C 312 | NP_006046.1 | |
XM_005246243.2 | 1010 | Missense Mutation | TAT,TGT | Y,C 324 | XP_005246300.1 |
LANCL1-AS1 - LANCL1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |