Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGAGCTGGGCAAGGGGAGCACGC[A/C]TCCGCAGCCCGGGGTGTCGCCACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138280 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLS - glutaminase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256310.1 | 530 | Missense Mutation | CAT,CCT | H,P 89 | NP_001243239.1 | |
NM_014905.4 | 530 | Missense Mutation | CAT,CCT | H,P 89 | NP_055720.3 | |
XM_005246467.3 | 530 | Missense Mutation | CAT,CCT | H,P 89 | XP_005246524.1 | |
XM_006712435.1 | 530 | Missense Mutation | CAT,CCT | H,P 89 | XP_006712498.1 | |
XM_017003829.1 | 530 | Missense Mutation | CAT,CCT | H,P 89 | XP_016859318.1 |