Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGTGTGTTTTATAGAATTCCCCT[C/G]TGTGCCCTGATAAATCAGCACCTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611678 | ||||||||||||||||||||
Literature Links: |
PDCL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDCL3 - phosducin like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024065.4 | 619 | Intron | NP_076970.1 | |||
XM_017004890.1 | 619 | Silent Mutation | CTC,CTG | L,L 76 | XP_016860379.1 |