Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGCAGGCGCCTCGTGGTTAGTTT[A/G]GGCACTGTTGATGTACTGAAGCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CYP20A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYP20A1 - cytochrome P450 family 20 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177538.2 | 865 | Silent Mutation | TTA,TTG | L,L 81 | NP_803882.1 | |
XM_005246698.3 | 865 | Intron | XP_005246755.1 | |||
XM_005246699.3 | 865 | Intron | XP_005246756.1 | |||
XM_005246701.3 | 865 | Intron | XP_005246758.1 | |||
XM_006712644.3 | 865 | Silent Mutation | TTA,TTG | L,L 63 | XP_006712707.1 | |
XM_006712645.3 | 865 | Silent Mutation | TTA,TTG | L,L 81 | XP_006712708.1 | |
XM_017004555.1 | 865 | Intron | XP_016860044.1 |