Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCTTCCGCACCTCCAAGTACCT[G/T]CTGGAGGAGTGGTTCCAGAACTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC442028 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC442028 - uncharacterized LOC442028 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEKT4 - tektin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286559.1 | Intron | NP_001273488.1 | ||||
NM_144705.3 | Intron | NP_653306.1 | ||||
XM_005263876.2 | Intron | XP_005263933.1 | ||||
XM_011510670.1 | Intron | XP_011508972.1 |