Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCTTTTGGTATCTAACCAGTGC[C/G]AACATCCCCAGTGAAACTTTAAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf47 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2orf47 - chromosome 2 open reading frame 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024520.2 | 1021 | Silent Mutation | GCC,GCG | A,A 233 | NP_078796.2 | |
XM_005246852.2 | 1021 | Intron | XP_005246909.1 |
TYW5 - tRNA-yW synthesizing protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |