Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCATTTGAGAAACATTTAATAAC[C/T]AAGATTTCCTATTTGAAAGTTCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610578 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP15 - Rho GTPase activating protein 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018460.3 | Intron | NP_060930.3 | ||||
XM_011511479.2 | Intron | XP_011509781.1 | ||||
XM_011511481.2 | Intron | XP_011509783.1 | ||||
XM_011511482.2 | Intron | XP_011509784.1 | ||||
XM_011511483.1 | Intron | XP_011509785.1 | ||||
XM_011511484.1 | Intron | XP_011509786.1 | ||||
XM_017004499.1 | Intron | XP_016859988.1 | ||||
XM_017004500.1 | Intron | XP_016859989.1 | ||||
XM_017004501.1 | Intron | XP_016859990.1 |