Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAAAAGTAGAGAGGTATGGGAGC[A/G]CAAAAGAAGATACTATTTTACTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607701 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FASTKD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FASTKD1 - FAST kinase domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281476.2 | Intron | NP_001268405.1 | ||||
NM_001322046.1 | Intron | NP_001308975.1 | ||||
NM_001322048.1 | Intron | NP_001308977.1 | ||||
NM_001322049.1 | Intron | NP_001308978.1 | ||||
NM_024622.5 | Intron | NP_078898.3 | ||||
XM_006712750.3 | Intron | XP_006712813.1 | ||||
XM_006712751.3 | Intron | XP_006712814.1 | ||||
XM_017004911.1 | Intron | XP_016860400.1 | ||||
XM_017004912.1 | Intron | XP_016860401.1 |
KLHL41 - kelch like family member 41 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |