Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTTTCTGTATTTAAGGAAGACA[C/T]AGTATAGCTTCCACAAGGAATTGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
KIAA1841 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIAA1841 - KIAA1841 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005264605.3 | 561 | Silent Mutation | CAC,CAT | H,H 115 | XP_005264662.1 | |
XM_005264606.4 | 561 | Silent Mutation | CAC,CAT | H,H 115 | XP_005264663.1 | |
XM_005264607.2 | 561 | Intron | XP_005264664.1 | |||
XM_011533126.2 | 561 | Silent Mutation | CAC,CAT | H,H 115 | XP_011531428.1 | |
XM_011533127.2 | 561 | Silent Mutation | CAC,CAT | H,H 115 | XP_011531429.1 | |
XM_011533128.1 | 561 | Silent Mutation | CAC,CAT | H,H 115 | XP_011531430.1 | |
XM_011533129.2 | 561 | Silent Mutation | CAC,CAT | H,H 115 | XP_011531431.1 | |
XM_011533131.2 | 561 | Silent Mutation | CAC,CAT | H,H 115 | XP_011531433.1 | |
XM_017005108.1 | 561 | Silent Mutation | CAC,CAT | H,H 115 | XP_016860597.1 | |
XM_017005109.1 | 561 | Silent Mutation | CAC,CAT | H,H 115 | XP_016860598.1 |