Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTTCTTCTCTCGGGACTCCTGCT[A/G]CCTCTGTGCCGCGCCTTCAACCTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 193210 | ||||||||||||||||||||
Literature Links: |
ITGAV PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITGAV - integrin subunit alpha V | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144999.2 | 351 | Intron | NP_001138471.1 | |||
NM_001145000.2 | 351 | Silent Mutation | CTA,CTG | L,L 25 | NP_001138472.1 | |
NM_002210.4 | 351 | Silent Mutation | CTA,CTG | L,L 25 | NP_002201.1 |