Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCAGGCCCAGGAAGGTGCTCTTG[A/G]AACCAAGATTCATAAACTAAGGCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611504 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CENPO - centromere protein O | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199803.2 | 620 | Missense Mutation | GAA,GGA | E,G 43 | NP_001186732.1 | |
NM_001322101.1 | 620 | Missense Mutation | GAA,GGA | E,G 49 | NP_001309030.1 | |
NM_024322.3 | 620 | Missense Mutation | GAA,GGA | E,G 49 | NP_077298.1 |
PTRHD1 - peptidyl-tRNA hydrolase domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |