Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCCCGTGCCTGGCCCAGGAGGG[C/T]CCCCAGGTACGTGTTGGCTCTCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 188855 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNLY PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GNLY - granulysin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302758.1 | 292 | Silent Mutation | GGC,GGT | G,G 77 | NP_001289687.1 | |
NM_006433.4 | 292 | Silent Mutation | GGC,GGT | G,G 50 | NP_006424.2 | |
NM_012483.3 | 292 | Silent Mutation | GGC,GGT | G,G 35 | NP_036615.2 | |
XM_005264084.2 | 292 | Silent Mutation | GGC,GGT | G,G 77 | XP_005264141.1 | |
XM_005264085.2 | 292 | Silent Mutation | GGC,GGT | G,G 50 | XP_005264142.1 | |
XM_005264087.2 | 292 | Silent Mutation | GGC,GGT | G,G 35 | XP_005264144.1 |