Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCACCAATATATTCAAACAAGAA[A/G]GAAAGCGATTTTAACACTCCATTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605590 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SF3B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SF3B1 - splicing factor 3b subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005526.2 | 3550 | Intron | NP_001005526.1 | |||
NM_001308824.1 | 3550 | Intron | NP_001295753.1 | |||
NM_012433.3 | 3550 | Silent Mutation | TCC,TCT | S,S 1152 | NP_036565.2 |