Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGGGAGTTGGCCTGCCTCGTGGC[C/T]GTGGGGGAGGGCAGGCAAGGCGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613817 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCTD18 - potassium channel tetramerization domain containing 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321547.1 | 1727 | Silent Mutation | ACA,ACG | T,T 395 | NP_001308476.1 | |
NM_001321548.1 | 1727 | Silent Mutation | ACA,ACG | T,T 186 | NP_001308477.1 | |
NM_001321550.1 | 1727 | Silent Mutation | ACA,ACG | T,T 186 | NP_001308479.1 | |
NM_152387.3 | 1727 | Silent Mutation | ACA,ACG | T,T 395 | NP_689600.2 |
SPATS2L - spermatogenesis associated serine rich 2 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |