Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGTTCGGGCCATGTCCTCGATCT[A/G]GGGGTAGGCCCCCAGGGCCTGGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607877 | ||||||||||||||||||||
Literature Links: |
COPS9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COPS9 - COP9 signalosome subunit 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OTOS - otospiralin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_148961.3 | 293 | Nonsense Mutation | CAG,TAG | Q,* 62 | NP_683764.1 | |
XM_017003409.1 | 293 | Nonsense Mutation | CAG,TAG | Q,* 81 | XP_016858898.1 | |
XM_017003410.1 | 293 | Nonsense Mutation | CAG,TAG | Q,* 62 | XP_016858899.1 |