Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACGTCCTACACTTGAGGGGTGT[G/T]TTGACTACCCAGCCTGTGGAAGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609803 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKAR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKAR - ankyrin and armadillo repeat containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ASNSD1 - asparagine synthetase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019048.2 | 1127 | Silent Mutation | GTG,GTT | V,V 59 | NP_061921.1 | |
XM_017004381.1 | 1127 | Silent Mutation | GTG,GTT | V,V 59 | XP_016859870.1 | |
XM_017004382.1 | 1127 | Silent Mutation | GTG,GTT | V,V 59 | XP_016859871.1 |